Appearance (Color)White to almost white or colorless
Appearance (Form)Crystalline powder or crystals
Titration with HClO497.5 to 102.5 % (dried substance)
Solubility(5 % in water) Clear colorless
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.