Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 448160050
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 448160050
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 448160050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10039-32-4
IUPAC Name
disodium dodecahydrate hydrogen phosphate
Molecular Formula
H25Na2O16P
InChI Key
DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
Titration with HClO4
>=98.5 %
Heavy metals (as Pb)
=<10 ppm
Appearance (Color)
White
Appearance (Form)
Crystals
Water
57 to 61 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text