Thermo Scientific Chemicals

N,N-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: 448251000
100 mL, Each
Thermo Scientific Chemicals

N,N-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: 448251000
100 mL, Each
Quantity
Catalog number: 448251000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
121-05-1
IUPAC Name
(2-aminoethyl)bis(propan-2-yl)amine
Molecular Formula
C8H20N2
InChI Key
CURJNMSGPBXOGK-UHFFFAOYSA-N
SMILES
CC(C)N(CCN)C(C)C
Refractive index
1.4420 to 1.4440 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text