Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Clear GPLE Reaction Plates with Barcode

Catalog number: 4483348
Have Questions?
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Clear GPLE Reaction Plates with Barcode

Catalog number: 4483348
Have Questions?
Catalog Number
4483348
Unit Size
20 plates
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
448334820 plates
-
4483351500 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems™ MicroAmp™ EnduraPlate™ GPLE plastic consumables provide superior PCR and qPCR performance and quality in formats developed to meet your experimental needs. All EnduraPlate™ GPLE plastic consumables are validated on Applied Biosystems™ thermal cyclers, real-time thermal cyclers, and genetic analyzers for optimal fit and performance.



Key Features and Benefits:



• Each lot is certified DNA, RNAse, and PCR inhibitor-free

• Labeled 'For Laboratory Use'

• Optimized for performance with Applied Biosystems™ instruments

• Excellent mechanical stability and flatness for your complete workflow needs



Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate offers mechanical stability, flatness, and excellent durability. ANSI/SBS-compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler → real-time thermal cycler → genetic analyzer).



EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black, well-identification text helps to minimize pipetting mistakes and improves sample preparation productivity.

Note: An adapter (Cat No. A24820) is required for use of this product with 7300/7500 Real-Time PCR systems.

Specifications

Barcode
Includes Barcode
Certifications/Compliance
Certified DNA/RNase Free
Packaging
20 Plates
Product Line
MicroAmp
Color
Optical, Clear
Product Type
Microplate
For Use With (Equipment)
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style
Semi
Material
Polycarbonate, Polypropylene
Volume (Metric) Well
0.3 mL
No. of Wells
96
Volume (Metric) Working
0.2 mL
Quantity
20 Plates
Reaction Speed
Fast, Standard

Contents & Storage

20 96-Well plates. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text