Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Red Reaction Plates with Barcode

Catalog number: 4483350
Have Questions?
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Red Reaction Plates with Barcode

Catalog number: 4483350
Have Questions?
Catalog Number
4483350
Unit Size
20 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
448335020 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. All of our plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. With the new EnduraPlate™ reaction plates, there is now a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments.

Key Features and Benefits:

• Optimized for performance with Applied Biosystems™ instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs

Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability. ANSI/SBS compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler -> real-time thermal cycler -> genetic analyzer).

EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity, while five color choices provide a convenient way to organize your laboratory assays.

Note: An adapter (Cat No. A24820) is required for use of this product with 7300/7500 Real-Time PCR systems.

Specifications

Barcode
Includes Barcode
Certifications/Compliance
Certified DNA/RNase Free
Packaging
20 Plates
Product Line
MicroAmp
Color
Optical, Red
Product Type
Microplate
For Use With (Equipment)
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style
Semi
Material
Polycarbonate, Polypropylene
Volume (Metric) Well
0.3 mL
No. of Wells
96
Volume (Metric) Working
0.2 mL
Reaction Speed
Standard

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text