Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
4 x 25 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 448391000
Price (USD)/ Each
72.65
Online exclusive
80.50 
Save 7.85 (10%)
-
Add to cart
Quantity:
4 x 25 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.50 
Save 7.85 (10%)
Add to cart
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals
Catalog number448391000
Price (USD)/ Each
72.65
Online exclusive
80.50 
Save 7.85 (10%)
-
Add to cart
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Acidity (CH3COOH)=<0.002 %
Residue after evaporation=<0.0005 %
Appearance (Color)Clear colorless
GC>=99.85 %
Water=<0.001 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only