Thermo Scientific Chemicals

Methanol, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals

Catalog number: 448411000
100 mL, Case of
Thermo Scientific Chemicals

Methanol, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, Thermo Scientific Chemicals

Catalog number: 448411000
100 mL, Case of
Quantity
Catalog number: 448411000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-56-1
IUPAC Name
methanol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES
CO
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
GC
>=99.85 %
Residue after evaporation
=<0.0005 %
Acidity/alkalinity
=<0.0005 meq/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text