Thermo Scientific Chemicals

Potassium trimethylsilanolate, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 448468000
800 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Potassium trimethylsilanolate, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 448468000
800 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 448468000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10519-96-7
IUPAC Name
trimethylsilanol potassium
Molecular Formula
C3H10KOSi
InChI Key
COTHYYYVPUZALV-UHFFFAOYSA-N
SMILES
[K].C[Si](C)(C)O
Appearance (Form)
Solution
Appearance (Color)
Colorless to straw yellow
Titration with HClO4
1.9 to 2.2 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text