Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals
Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Catalog number 448515000
Price (USD)/ Each
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
Molecular Weight (g/mol)37.83
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Iron (Fe)=<10 ppm
Methanol=<2000 ppm
Appearance (Form)Clear liquid
Titration with HCl38.0 to 42.0 % w/w (NaOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only