Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 448540050
Price (USD)/ Each
79.65
Online exclusive
93.10 
Save 13.45 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
93.10 
Save 13.45 (14%)
Add to cart
Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Catalog number448540050
Price (USD)/ Each
79.65
Online exclusive
93.10 
Save 13.45 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namediammonium dioxomolybdenumbis(olate)
Molecular FormulaH8MoN2O4
InChI KeyAPUPEJJSWDHEBO-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].[O-][Mo]([O-])(=O)=O
Molecular Weight (g/mol)196.02
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Total trace metal impurities=<100 ppm
Appearance (Form)Powder or crystals and/or chunks
Molybdenum (Mo)47.0 to 50.7 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only