Thermo Scientific Chemicals

Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 448541000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 448541000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 448541000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13106-76-8
IUPAC Name
diammonium dioxomolybdenumbis(olate)
Molecular Formula
H8MoN2O4
InChI Key
APUPEJJSWDHEBO-UHFFFAOYSA-P
SMILES
[NH4+].[NH4+].[O-][Mo]([O-])(=O)=O
Appearance (Form)
Powder or crystals and/or chunks
Appearance (Color)
White to light yellow
Total trace metal impurities
=<100 ppm
Molybdenum (Mo)
47.0 to 50.7 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text