Applied Biosystems™

QuantStudio™ 7 Flex Real-Time PCR System, 96-well, desktop

Catalog number: 4485690
Have Questions?
Applied Biosystems™

QuantStudio™ 7 Flex Real-Time PCR System, 96-well, desktop

Catalog number: 4485690
Have Questions?
Catalog Number
4485690
Unit Size
1 system
Price (USD)
Catalog NumberUnit SizePrice (USD)
44856901 systemRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems® QuantStudio™ 7 Flex Real-Time PCR System advances your research further by enabling a broad range of real-time PCR-based applications through its multiplexing capabilities and interchangeable block formats. With a simplified workflow, intuitive software, automation capabilities, and touch-screen interface, the QuantStudio™ 7 Flex system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.

Features of the QuantStudio™ 7 Flex Real-Time PCR System include:

• Increased application versatility through the accomodation of 4 different block types
• Compatible with over 8 million TaqMan® assays for gene expression, genetic variation, gene regulation, or protein expression experiments
• The intelligent design of the QuantStudio™ platform combined with Applied Biosystems® instrument reliability and accuracy
• Easy-to-use software, responsive touch-screen, automation capabilities, and effortless block exchange without the need for any tools

Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

>> Download a 90-day trial of the QuantStudio™ 7 software

The QuantStudio™ 7 Flex system accommodates the interchange of a 96-well, 96-well Fast, 384-well, or TaqMan® array card block. The system is delivered with one block type (a 96-well block in this case); other block types may be purchased separately. In addition, a Twister® Robot Automation Accessory is available for this system to support high throughput workflows. For a more value-priced system that does not support TaqMan® array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio™ 6 Flex system. And for the ultimate in flexibility, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan® OpenArray® plates.

Accomplish More, Faster
Run hundreds of real-time PCR reactions effortlessly using TaqMan® Array 384-well microfluidic cards. And when integrated with the Twister® Robot Automation Accessory (available separately), the QuantStudio™ 7 Flex system maximizes your throughput capabilities.

High Application Versatility
Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, High Resolution Melt (HRM), and Protein Thermal Shift™ experiments.

Results You Can Trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex™ system, which features 6 decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.

Sensitive Data Analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Block Format
Interchangeable
Format
96-well Plate
For Use With (Equipment)
QuantStudio™
Product Line
QuantStudio™
Display Type
Touchscreen
Quantity
1 system
Unit Size
1 system

Contents & Storage

Desktop Computer

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text