Applied Biosystems™

QuantStudio™ 6 Flex Real-Time PCR System, 96-well, desktop

Catalog number: 4485692
Have Questions?
Applied Biosystems™

QuantStudio™ 6 Flex Real-Time PCR System, 96-well, desktop

Catalog number: 4485692
Have Questions?
Catalog Number
4485692
Unit Size
1 system
Price (USD)
Catalog NumberUnit SizePrice (USD)
44856921 systemRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems® QuantStudio™ 6 Flex Real-Time PCR System facilitates real-time PCR based applications with an upgradable block selection, allowing you to plan for experiments today and grow with tomorrow’s needs. With a simplified workflow, intuitive software, and touch-screen interface, the QuantStudio™ 6 Flex system offers exceptional reproducibility at a price you can afford.

Features of the QuantStudio™ 6 Flex Real-Time PCR System include:

• Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
• Compatible with over 8 million TaqMan® assays for gene expression, genetic variation, gene regulation, or protein expression experiments
• The intelligent design of the QuantStudio™ platform combined with Applied Biosystems® instrument reliability and accuracy
• Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools

Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

>> Download a 90-day trial of the QuantStudio™ 6 software

The QuantStudio™ 6 Flex system accommodates the interchange of a 96-well, 96-well Fast, or 384-well block. The system is delivered with one block type (a 96-well block in this case); other block types may be purchased separately. For a system that also supports TaqMan® array cards, as well as automation, please see our QuantStudio™ 7 Flex system. The QuantStudio™ 6 Flex system can also be upgraded to a QuantStudio™ 7 Flex system at a later date, as your needs change. For the ultimate in productivity, consider the QuantStudio™ 12K Flex system, which also accommodates TaqMan® OpenArray® plates.

Upgrade Capabilities When You Need Them

The QuantStudio™ 6 Flex system can be seamlessly upgraded to a QuantStudio™ 7 Flex system by a field service engineer to give you additional automation, throughput, and multiplexing capabilities.

Results You Can Trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex® system, which features 5 coupled excitation and emission filter channels.

Sensitive Data Analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Block Format
Interchangeable
Format
96-well Plate
For Use With (Equipment)
QuantStudio™
Product Line
QuantStudio™
Display Type
Touchscreen
Description
QuantStudio™ 6 Flex Real Time PCR System, 96-well
Quantity
1 system

Contents & Storage

Desktop Computer

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text