4-Cyano-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
4-Cyano-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 448710010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS843663-18-3
IUPAC Name(4-cyano-3-fluorophenyl)boronic acid
Molecular FormulaC7H5BFNO2
InChI KeyDECWLXUOZUMPBF-UHFFFAOYSA-N
SMILESOB(O)C1=CC(F)=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow
Titration with NaOH>=96.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only