Thermo Scientific Chemicals

2-Bromo-4-chloro-1-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 448760250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromo-4-chloro-1-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 448760250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 448760250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
31928-44-6
IUPAC Name
2-bromo-4-chloro-1-iodobenzene
Molecular Formula
C6H3BrClI
InChI Key
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
SMILES
ClC1=CC(Br)=C(I)C=C1
Appearance (Color)
Orange to brown
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text