Thermo Scientific Chemicals

5-Methoxyisatin, 97%, Thermo Scientific Chemicals

Catalog number: 448830250
25 g, Each
Thermo Scientific Chemicals

5-Methoxyisatin, 97%, Thermo Scientific Chemicals

Catalog number: 448830250
25 g, Each
Quantity
Catalog number: 448830250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39755-95-8
IUPAC Name
5-methoxy-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C9H7NO3
InChI Key
DMHGXMPXHPOXBF-UHFFFAOYSA-N
SMILES
COC1=CC=C2NC(=O)C(=O)C2=C1
Appearance (Color)
Dark red to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
199°C to 202°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text