Applied Biosystems™

TaqMan™ OpenArray™ PGx Express Panel, QuantStudio™ 12K Flex

Catalog number: 4488847
Applied Biosystems™

TaqMan™ OpenArray™ PGx Express Panel, QuantStudio™ 12K Flex

Catalog number: 4488847
Catalog Number
4488847
Unit Size
1 each
Price (HKD)
Price: 8,787.00
Your price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (HKD)AvailabilityQuantity
44888471 each
Price: 8,787.00
Your price:
***
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The TaqMan™ OpenArray™ PGx (Pharmacogenomics) Express Panel is an efficient, easy-to-use OpenArray™ plate for pharmacogenomics applications. The assays were developed to detect polymorphisms in genes encoding drug metabolism enzymes (DMEs) and associated transport proteins. This pharmacogenomics express panel contains 60 TaqMan™ DME and SNP genotyping assays. These assays provide coverage of essential, commonly studied markers within CYP2D6, CYP2C9, CYP2C19, and other important DME and clinical research genes.

Each TaqMan™ Drug Metabolism and SNP genotyping assay in the panel contains two allele-specific probes and a primer pair to detect the specific SNP target. Both the probes and primers uniquely align within the genome, enabling the TaqMan™ genotyping technology to provide superior specificity. It is this specificity that allows these assays to detect targets residing in highly homologous gene families that may include pseudogenes. TaqMan™ Drug Metabolism genotyping assays were developed using a high level of bioinformatics and wet-lab stringency. All assays have passed performance tests involving 180 unique DNA samples from four different populations.

The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.

For fast delivery, all assays in this collection have been manufactured and placed into inventory and are ready to ship at ambient temperature.

This panel is intended for use with the QuantStudio™ 12K Flex Real-time PCR System, which can cycle and image 4 plates simultaneously, providing high-throughput results with ease.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
OpenArray Plate
GC-Rich PCR Performance
High
Reaction Speed
Fast
Label or Dye
FAM
Product Line
TaqMan™
Product Type
Pgx Express Panel
Species
Human
For Use With (Application)
Genotyping

Contents & Storage

Includes one OpenArray™ Plate containing a defined panel of pre-loaded, bench-validated TaqMan™ assays.
Store at -5 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text