Thermo Scientific™
AminoLink™ Reductant
Catalog number: 44892
Have Questions?
Thermo Scientific™
AminoLink™ Reductant
Catalog number: 44892
Have Questions?
Catalog Number
44892
Unit Size
2 x 1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
448922 x 1 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific AminoLink Reductant (sodium cyanoborohydride) promotes the formation of stable bonds between aldehyde- and amine-containing molecules, enabling efficient labeling, conjugation, and immobilization of proteins and other molecules. Carbonyl groups such as aldehydes, ketones, and glyoxals react spontaneously with amino groups to form Schiff base intermediates that are in equilibrium with their free forms. The interaction is pH-dependent, being somewhat more efficient in acidic conditions, and especially strong at high pH. Addition of AminoLink Reductant to a reaction in which Schiff base formation has occurred results in complete reduction of the labile Schiff base intermediate to a chemically stable bond. Unlike sodium borohydride, sodium cyanoborohydride is sufficiently mild to avoid adversely reducing aldehydes to nonreactive hydroxyls.
The overall reaction of carbonyl groups to primary amino groups using sodium cyanoborohydride is called reductive amination. Immobilization by reductive amination of amine-containing biological molecules onto aldehyde-containing solid supports is used to create matrices for affinity purification of antibodies and other molecules. To conjugate glycoproteins by reductive amination, sugars in the polysaccharide groups must be oxidized using sodium meta-periodate to create reactive aldehyde groups. Treatment of glycoproteins with sodium meta-periodate generally creates sufficient aldehyde groups to be used as targets for efficient conjugation with amine- or hydrazide-containing molecules, including other proteins, biotin reagents and fluorescent labels. Because glycoproteins contain their own primary amino groups, polymerization of an oxidized protein may occur unless a sufficient molar excess of the intended amine-containing target molecule is added to the reaction.
Related Products
AminoLink™ Coupling Resin
AminoLink™ Immobilization Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Form
Powder
Product Line
AminoLink™
Type
Resin
Contents & Storage
Store desiccated at room temperature. Shipped at ambient temperature