Chemical Identifiers
CAS330645-87-9
IUPAC Namehexafluoro-λ⁵-phosphanuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
Molecular FormulaC11H15ClF6N5OP
InChI KeyZHHGTMQHUWDEJF-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C
View more