Pierce™ IgM Purification Kit
Pierce™ IgM Purification Kit
Thermo Scientific™

Pierce™ IgM Purification Kit

The Thermo Scientific Pierce IgM Purification Kit is an easy-to-use affinity matrix and buffer system to purify IgM from mouseRead more
Have Questions?
Catalog NumberQuantity
448971 Column
Catalog number 44897
Price (USD)
964.65
Online Exclusive
1,164.00
Save 199.35 (17%)
Each
Add to cart
Quantity:
1 Column
Request bulk or custom format
Price (USD)
964.65
Online Exclusive
1,164.00
Save 199.35 (17%)
Each
Add to cart
The Thermo Scientific Pierce IgM Purification Kit is an easy-to-use affinity matrix and buffer system to purify IgM from mouse ascites fluid; the method uses immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.

Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5 mL column of the MBP Agarose.

Features of the IgM purification kit:

Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum
Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5 mg IgM from 0.5 mL ascites samples per column-use
Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies
Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity

Mannan binding protein (MBP), also known as mannose-binding lectin (MBL) is a mammalian lectin that is produced in the liver and occurs in serum. MBP consists of 18 identical subunits, each with a molecular mass of approximately 31kDa. The protein is capable of initiating carbohydrate-mediated complement activation.

The IgM Purification Kit protocol based on immobilized MBP is most effective for purifying mouse IgM from ascites fluid, although it also enables partial purification of human IgM from serum. Binding and elution steps are temperature- and calcium-dependent. Binding and washing steps are performed at 4°C in Tris buffer that contains 20 mM calcium chloride. Elution is achieved at room temperature in Tris buffer that contains EDTA but no calcium chloride. The simple protocol is easy to use and yields 90% pure mouse IgM from ascites. Immobilized MBP can be regenerated at least 10 times with no apparent loss of binding capacity. Human IgM also will bind to the support but with slightly lower capacity and some copurification of IgG. The purification system has not been tested or validated for use with other species and sources of samples.

Related Products
Pierce™ Mannan Binding Protein Agarose
MBP Column Preparation Buffer for Pierce™ IgM Purification Kit
Elution Buffer for Pierce™ IgM Purification Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce IgM Purification Kit
Purification TargetIgM Antibody
Quantity1 Column
Column TypeAgarose Resin, Affinity
FormatKit
Product LinePierce™
Product TypeIgM Purification Kit
Stationary PhaseLectin
Unit SizeEach
Contents & Storage
Sufficient For: 10 IgM purifications, each from 0.5 mL mouse ascites samples
• MBP Agarose Column, 5 mL, 1 column
• IgM Binding Buffer, 800 mL
• IgM Elution Buffer, 500 mL
• MBP Column Preparation Buffer, 50 mL
• Column Extender

Store at 4°C

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link