Thermo Scientific™

CarboxyLink™ Immobilization Kit, 2 mL

Catalog number: 44899
Have Questions?
Thermo Scientific™

CarboxyLink™ Immobilization Kit, 2 mL

Catalog number: 44899
Have Questions?
Catalog Number
44899
Unit Size
5 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
448995 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific CarboxyLink Immobilization Kit is for covalent immobilization of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.

CarboxyLink Resin is crosslinked beaded agarose that has been activated with diamino-dipropylamine (DADPA) to contain long spacer arms, each with a primary amine at the end. When incubated with the resin and the carbodiimide crosslinker EDC (included in the CarboxyLink Immobilization Kit), carboxyl-containing molecules become permanently attached to the support by stable amide bonds. Once a molecule is coupled, the prepared affinity column can be used multiple times in typical protein affinity purification procedures. CarboxyLink Coupling Resins can also be used to immobilize other kinds of molecules using alternative amine-reactive crosslinking chemistries.

Features of the CarboxyLink Immobilization Kit:

CarboxyLink Coupling Resin—DADPA-activated crosslinked 4% beaded agarose
Efficient immobilization—Couple 1 to 2 mg of peptide per milliliter of resin (CarboxyLink Agarose Resin activated with greater than 16 μMol amine milliliter of resin; DADPA on UltraLink Support activated with greater than 40 μMol amine milliliter of resin)
Stable linkage—immoblization results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin
Flexible and gentle coupling conditions—immobilization reaction completed in simple MES or other non-amine and non-carboxyl, near-neutral buffer, with or without organic solvent.
Ideal for unmodified peptides—immobilizes peptides with high capacity and various orientations without steric hindrance, allowing for effective use in affinity purification of specific antibodies
Convenient kits and product sizes—choose 5-column kits with complete sets of buffers, crosslinker and versatile spin/drip columns containing either type of resin (agarose or polyacrylamide) or choose stand-alone resin for other uses.

Because EDC crosslinks carboxylates to primary amines, peptides and other molecules that contain both of these functional groups will be variously polymerized as well as immobilized to the CarboxyLink Resin. For peptides whose only carboxyl and amino groups are at the termini, coupling with the CarboxyLink Kit will result in end-to-end chains of peptide that are linked by their C-terminal ends to the support. Generally, this structure provides for high capacity and low steric hindrance for use in affinity purification. However, in the case of peptides containing multiple primary amines (lysine) or carboxylates (glutamic and aspartic acid) in the middle of their sequence, the CarboxyLink Coupling Method may cause key binding sites to be blocked, resulting in high coupling capacity but poor binding capacity for affinity purification. Alternative methods for immobilization include AminoLink Plus Coupling Resin, UltraLink Biosupport and SulfoLink Coupling Resin.

Related Products
CarboxyLink™ Coupling Resin
CarboxyLink™ Immobilization Kit with UltraLink™ Support, 2 mL
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
2 mL
Description
CarboxyLink Immobilization Kit, 2mL
Product Line
CarboxyLink™
Quantity
1 Kit
Target
Antibody
Type
Immobilization Kit
Stationary Phase
Amine
Sufficient For
5 Columns
Format
Spin Column
Column Type
Affinity, Agarose Resin

Contents & Storage

Sufficient For: Immobilizing 1 to 2 mg of peptide per column
• CarboxyLink Columns, 2 mL, 5 columns
• EDC Crosslinker, 5 x 60 mg
• MES Buffered Saline Pack (makes 500 mL), 1 pack
• Wash Solution, 120 mL
• Column Accessories

Store EDC desiccated at -20°C. Store other components at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text