1x1 image pixel for data collection
(-)-Epigallocatechin gallate, 95%
(-)-Epigallocatechin gallate, 95%
(-)-Epigallocatechin gallate, 95%
(-)-Epigallocatechin gallate, 95%
Thermo Scientific Chemicals

(-)-Epigallocatechin gallate, 95%

CAS: 989-51-5 | C22H18O11 | 458.37 g/mol
Have Questions?
Catalog NumberQuantity
44901010010 mg
Catalog number 449010100
Price (USD)
39.65
Online Exclusive
44.00
Save 4.35 (10%)
Each
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.00
Save 4.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS989-51-5
IUPAC Name(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Molecular FormulaC22H18O11
InChI KeyWMBWREPUVVBILR-WIYYLYMNSA-N
SMILESOC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@H](OC2=C1)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to light pink
HPLC>=94.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link