Thermo Scientific Chemicals

(-)-Epigallocatechin gallate, 95%

Catalog number: 449010500
50 mg, Each
Thermo Scientific Chemicals

(-)-Epigallocatechin gallate, 95%

Catalog number: 449010500
50 mg, Each
Quantity
Catalog number: 449010500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
989-51-5
IUPAC Name
(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Molecular Formula
C22H18O11
InChI Key
WMBWREPUVVBILR-WIYYLYMNSA-N
SMILES
OC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@H](OC2=C1)C1=CC(O)=C(O)C(O)=C1
Appearance (Color)
White to light pink
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=94.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text