Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethanolamine, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 449062500
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
Add to cart
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Catalog number449062500
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
-
Add to cart
Chemical Identifiers
CAS111-42-2
IUPAC Name2-[(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyZBCBWPMODOFKDW-UHFFFAOYSA-N
SMILESOCCNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
GC>=98.5 %
GC=<1.0 % (Triethanolamine)
Water=<0.15 %
Color scale=<15 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only