Thermo Scientific™

Pierce™ Thiophilic Adsorption Kit

Catalog number: 44916
Have Questions?
Thermo Scientific™

Pierce™ Thiophilic Adsorption Kit

Catalog number: 44916
Have Questions?
Catalog Number
44916
Unit Size
4 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
449164 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Thiophilic Adsorbent is a protein-free immunosorbent resin for gentle preferential binding and purification of immunoglobulins from a variety of sources including mouse, rat, rabbit, goat and human. This Pierce Thiophilic Adsorption Kit is a purification kit containing columns pre-packed with the resin and ready-to-use buffers.

Features of Thiophilic Adsorbent:

High capacity – gently bind and purify approx. 20 mg polyclonal antibody per milliliter of adsorbent resin
Broad antibody specificity – effective for most immunoglobulin classes and isotypes derived from many different animal species, including chicken IgY
Simple and fast – quickly fractionate and purify the immunoglobulin component of serum or ascites samples without subsequent dialysis or desalting steps
Gentle – binding condition is nondenaturing and the elution buffer yields concentrated, salt-free immunoglobulin at near neutral pH; provides excellent recovery of functional antibody

Pierce Thiophilic Adsorbent is 6% beaded agarose that has been modified to contain simple sulfone-thioether groups. The result is an affinity resin that has a high binding capacity and a broad specificity for many species of immunoglobulins when incubated in 0.5M potassium sulfate. In effect, thiophilic adsorption is a resin-based variation of ammonium sulfate precipitation for quick immunoglobulin purification from crude serum samples. Unlike antibody fractionation by ammonium sulfate precipitation, purification with thiophilic adsorbent yields concentrated, essentially salt-free, highly purified immunoglobulins at near-neutral pH. Thus, this simple one-step method eliminates the need for additional treatment of the sample for storage or for subsequent conjugation reactions. The gentle binding and elution conditions ensure a high protein recovery with excellent preservation of antibody activity.

Thiophilic adsorption is based on the ability of some proteins, particularly immunoglobulins, to bind to an immobilized ligand that contains a sulfone group in close proximity to a thioether. Salts that interact with water molecules, such as potassium and ammonium sulfate, promote this binding by driving the protein into close proximity with the ligand.

Related Products
Pierce™ Thiophilic Adsorbent
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Adsorption Kit
Product Line
Pierce™
Column Type
Agarose Resin, Affinity
Format
Gravity Flow Column
Stationary Phase
Sulfone Thioether
Purification Target
IgG Antibody
Description
Pierce Thiophilic Adsorption Kit
Quantity
4 Columns

Contents & Storage

Sufficient For: Purifying up to 60 mg immunoglobulin per column
• Thiophilic Adsorbent Columns, 3 mL, 4 columns
• Binding Buffer, 1 L
• Elution Buffer, 1 L
• Column Storage Buffer (2X), 100 mL
• Guanidine-HCl Crystals, 230 g
• Column Accessories

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text