Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals

Catalog number: 449180050
5 L, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals

Catalog number: 449180050
5 L, Each
Quantity
Catalog number: 449180050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Color scale
=<100 APHA
Refractive index
1.4690 to 1.4710 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text