Thermo Scientific Chemicals

Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals

Catalog number: 449220250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals

Catalog number: 449220250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 449220250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-24-6
IUPAC Name
trimethylstannanol
Molecular Formula
C3H10OSn
InChI Key
OJZNYUDKNVNEMV-UHFFFAOYSA-M
SMILES
C[Sn](C)(C)O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
114°C to 122°C
Elemental Analysis
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text