Methyl hexanoate, 99%, Thermo Scientific Chemicals
Methyl hexanoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl hexanoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 449240500
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl hexanoate
Molecular FormulaC7H14O2
InChI KeyNUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILESCCCCCC(=O)OC
Molecular Weight (g/mol)130.19
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Colorless to light yellow
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only