1-Acetylpyrene, 97%, Thermo Scientific Chemicals
1-Acetylpyrene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Acetylpyrene, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 449300050
Price (USD)/ Each
120.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
120.00
Add to cart
1-Acetylpyrene, 97%, Thermo Scientific Chemicals
Catalog number449300050
Price (USD)/ Each
120.00
-
Add to cart
Chemical Identifiers
CAS3264-21-9
IUPAC Name1-(pyren-1-yl)ethan-1-one
Molecular FormulaC18H12O
InChI KeyKCIJNJVCFPSUBQ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Melting point86°C to 89°C
Infrared spectrumConforms
Appearance (Color)Yellow to light brown
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only