2,6-Dibromonaphthalene, 99%, Thermo Scientific Chemicals
2,6-Dibromonaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dibromonaphthalene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 449340050
Price (USD)/ Each
182.65
Online exclusive
203.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
182.65
Online exclusive
203.00 
Save 20.35 (10%)
Add to cart
2,6-Dibromonaphthalene, 99%, Thermo Scientific Chemicals
Catalog number449340050
Price (USD)/ Each
182.65
Online exclusive
203.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13720-06-4
IUPAC Name2,6-dibromonaphthalene
Molecular FormulaC10H6Br2
InChI KeyPJZDEYKZSZWFPX-UHFFFAOYSA-N
SMILESBrC1=CC2=CC=C(Br)C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Powder
Melting point162°C to 167°C
Infrared spectrumConforms
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only