4'-Chloro-2'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals
4'-Chloro-2'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Chloro-2'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 449360500
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-(4-chloro-2-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H7ClO2
InChI KeyQCVSDCHNBNFJDQ-UHFFFAOYSA-N
SMILESCC(=O)C1=C(O)C=C(Cl)C=C1
Molecular Weight (g/mol)170.59
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=96.0 %
Melting point27°C to 30°C
Appearance (Form)Crystalline mass
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only