Thermo Scientific Chemicals

(S)-Styrene oxide, 94%, Thermo Scientific Chemicals

Catalog number: 449450250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-Styrene oxide, 94%, Thermo Scientific Chemicals

Catalog number: 449450250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 449450250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20780-54-5
IUPAC Name
(2S)-2-phenyloxirane
Molecular Formula
C8H8O
InChI Key
AWMVMTVKBNGEAK-MRVPVSSYSA-N
SMILES
C1O[C@H]1C1=CC=CC=C1
Appearance (Form)
Liquid
Refractive index
1.5340 to 1.5360 (20°C, 589 nm)
Water
=<0.5 %
GC
>=92 %
Appearance (Color)
Colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text