Thermo Scientific Chemicals

Cytidine 5'-monophosphate, 97%, from yeast

Catalog number: 449490050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Cytidine 5'-monophosphate, 97%, from yeast

Catalog number: 449490050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 449490050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63-37-6
IUPAC Name
{[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Molecular Formula
C9H14N3O8P
InChI Key
IERHLVCPSMICTF-NHBCCFCANA-N
SMILES
NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]1O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Loss on drying
=<3 %
Appearance (Color)
White to off-white
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text