Thermo Scientific Chemicals

5-Hexenoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 449520050
5 g, Each
Thermo Scientific Chemicals

5-Hexenoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 449520050
5 g, Each
Quantity
Catalog number: 449520050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1577-22-6
IUPAC Name
hex-5-enoic acid
Molecular Formula
C6H10O2
InChI Key
XUDOZULIAWNMIU-UHFFFAOYSA-N
SMILES
OC(=O)CCCC=C
Infrared spectrum
Conforms
Appearance (Color)
Colorless to light yellow
Appearance (Form)
Liquid
Refractive index
1.4340 to 1.4360 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text