Thermo Scientific Chemicals

5-Acetylvaleric acid, 90+%, technical, Thermo Scientific Chemicals

Catalog number: 449530250
25 g, Each
Thermo Scientific Chemicals

5-Acetylvaleric acid, 90+%, technical, Thermo Scientific Chemicals

Catalog number: 449530250
25 g, Each
Quantity
Catalog number: 449530250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3128-07-2
IUPAC Name
6-oxoheptanoic acid
Molecular Formula
C7H12O3
InChI Key
IZOQMUVIDMLRDC-UHFFFAOYSA-N
SMILES
CC(=O)CCCCC(O)=O
Appearance (Color)
White to off-white
GC
>=90.0 %
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text