Thermo Scientific Chemicals

5-Amino-3-methyl-1-phenylpyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 449600250
25 g, Each
Thermo Scientific Chemicals

5-Amino-3-methyl-1-phenylpyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 449600250
25 g, Each
Quantity
Catalog number: 449600250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1131-18-6
IUPAC Name
3-methyl-1-phenyl-1H-pyrazol-5-amine
Molecular Formula
C10H11N3
InChI Key
FMKMKBLHMONXJM-UHFFFAOYSA-N
SMILES
CC1=NN(C(N)=C1)C1=CC=CC=C1
Melting point
114°C to 117°C
Appearance (Form)
Crystalline powder and/or chunks
Appearance (Color)
White to brown
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text