Potassium fluoride, Thermo Scientific Chemicals
Potassium fluoride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium fluoride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 449655000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium fluoride
Molecular FormulaFK
InChI KeyNROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES[F-].[K+]
Molecular Weight (g/mol)58.10
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=99.0 %
Potassium fluosilicate=<0.1 % (K2SiF6)
Heavy metals (as Pb)=<0.001 %
Appearance (Color)White
Titrable acid=<0.03 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only