4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals
4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals
4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals
4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4497700505 g
Catalog number 449770050
Price (USD)
90.65
Special offer
Online exclusive
Ends: 26-Sep-2025
111.00
Save 20.35 (18%)
Each
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 26-Sep-2025
111.00
Save 20.35 (18%)
Each
Add to cart
Chemical Identifiers
CAS16520-62-0
IUPAC Name(but-3-yn-1-yl)benzene
Molecular FormulaC10H10
InChI KeyQDEOKXOYHYUKMS-UHFFFAOYSA-N
SMILESC#CCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5210 to 1.5240 (20°C, 589 nm)
Appearance (Form)Liquid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link