Thermo Scientific Chemicals

1,2-Diamino-4,5-difluorobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 449810050
5 g, Each
Thermo Scientific Chemicals

1,2-Diamino-4,5-difluorobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 449810050
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 449810050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76179-40-3
IUPAC Name
4,5-difluorobenzene-1,2-diamine
Molecular Formula
C6H6F2N2
InChI Key
PPWRHKISAQTCCG-UHFFFAOYSA-N
SMILES
NC1=CC(F)=C(F)C=C1N
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Appearance (Color)
Off-white to red-brown
Appearance
May darken on storage
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text