6-Fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific Chemicals
6-Fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 449850250
Price (USD)/ Each
552.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
650.00 
Save 97.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
552.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
650.00 
Save 97.35 (15%)
Add to cart
6-Fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific Chemicals
Catalog number449850250
Price (USD)/ Each
552.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
650.00 
Save 97.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name6-fluoropyridine-3-carbaldehyde
Molecular FormulaC6H4FNO
InChI KeyPZPNGWWKCSJKOS-UHFFFAOYSA-N
SMILESFC1=NC=C(C=O)C=C1
Molecular Weight (g/mol)125.10
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to white to light yellow
GC>=94 %
Appearance (Form)Liquid or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only