Thermo Scientific Chemicals

3-Hydroxy-4-quinolinecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 449930010
1 g, Each
Thermo Scientific Chemicals

3-Hydroxy-4-quinolinecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 449930010
1 g, Each
Quantity
Catalog number: 449930010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-13-8
IUPAC Name
3-hydroxyquinoline-4-carboxylic acid
Molecular Formula
C10H7NO3
InChI Key
MXNVEJDRXSFZQB-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=CC=CC2=NC=C1O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Form)
Crystalline powder and/or chunks
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text