Thermo Scientific Chemicals

2-Chloro-4-ethynylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 449980010
1 g, Each
Thermo Scientific Chemicals

2-Chloro-4-ethynylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: 449980010
1 g, Each
Quantity
Catalog number: 449980010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
945717-09-9
IUPAC Name
2-chloro-4-ethynylpyridine
Molecular Formula
C7H4ClN
InChI Key
DLEWWBKXDRBGRF-UHFFFAOYSA-N
SMILES
ClC1=NC=CC(=C1)C#C
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
White to light brown to green
HPLC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text