Thermo Scientific Chemicals

4-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: 450000010
1 g, Each
Thermo Scientific Chemicals

4-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: 450000010
1 g, Each
Quantity
Catalog number: 450000010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100367-39-3
IUPAC Name
4-bromo-2-methoxypyridine
Molecular Formula
C6H6BrNO
InChI Key
YFTGMMXMLPTTAY-UHFFFAOYSA-N
SMILES
COC1=NC=CC(Br)=C1
Infrared spectrum
Conforms
Refractive index
1.5540 to 1.5560 (20°C, 589 nm)
Appearance (Color)
Colorless to yellow
Appearance (Form)
Low melting solid or liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text