Specifications
3'Primer ModificationTAMRA
5'Primer ModificationFAM, TET, VIC
FormLiquid or Dried Down
FormatTube
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Green FeaturesSustainable packaging
Internal Probe ModificationNone
Product LineTaqMan™
Product TypeTAMRA Probe
Purification MethodHPLC
Purity or Quality GradeHPLC
Shipping ConditionRoom Temperature
For Use With (Application)Real Time PCR (qPCR)
Unit Size6,000 pmol