Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling, for mass spectrometry, Thermo Scientific Chemicals
Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling, for mass spectrometry, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling, for mass spectrometry, Thermo Scientific Chemicals
Have Questions?
Quantity:
1 mL
Packaging:
Glass bottle
Catalog number 450130010
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
336.00 
Save 50.35 (15%)
-
Add to cart
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
336.00 
Save 50.35 (15%)
Add to cart
Perfluorokerosene (PFK), high boiling, 50% in PFK low boiling, for mass spectrometry, Thermo Scientific Chemicals
Catalog number450130010
Price (USD)/ Each
285.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
336.00 
Save 50.35 (15%)
-
Add to cart
Chemical Identifiers
CASna
SpecificationsSpecification SheetSpecification Sheet
Suitability(for mass analysis) Passes test
Appearance (Form)Liquid
Appearance (Color)Clear colorless