Thermo Scientific Chemicals

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000 to 180,000, Thermo Scientific Chemicals

Catalog number: 450155000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000 to 180,000, Thermo Scientific Chemicals

Catalog number: 450155000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 450155000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9002-89-5
Molecular Formula
(C2H4O)n
SMILES
OC(-*)C-*
MDL Number
MFCD00081922
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Total volatile
=<5 % (including water)
Appearance (Form)
Granular solid
pH
4.5 to 6.5 (4 % in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text