4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers
4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers
4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers
4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers
Thermo Scientific Chemicals

4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers

CAS: 6864-37-5 | C15H30N2 | 238.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 450242500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6864-37-5
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4980 to 1.5000 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0446558Certificate of AnalysisMay 17, 2025450242500, 450240010
A0459649Certificate of AnalysisJan 30, 2024450242500, 450240010
A0448651Certificate of AnalysisDec 30, 2022450242500, 450240010
A0393725Certificate of AnalysisDec 14, 2022450242500, 450240010
A0415587Certificate of AnalysisDec 12, 2022450242500, 450240010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link