Thermo Scientific Chemicals

4'-Chlorobiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 450330100
10 g, Each
Thermo Scientific Chemicals

4'-Chlorobiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 450330100
10 g, Each
Quantity
Catalog number: 450330100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20443-74-7
IUPAC Name
4'-chloro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Formula
C12H8Cl2O2S
InChI Key
NWYUSJMIHFIMTA-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Appearance (Color)
White to off-white
Melting point
106°C to 112°C
Appearance (Form)
Powder
HPLC
>=96 % as diethylsulfonamide
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text