4'-Chlorobiphenyl-4-carboxaldehyde, 97%
4'-Chlorobiphenyl-4-carboxaldehyde, 97%
4'-Chlorobiphenyl-4-carboxaldehyde, 97%
4'-Chlorobiphenyl-4-carboxaldehyde, 97%
Thermo Scientific Chemicals

4'-Chlorobiphenyl-4-carboxaldehyde, 97%

CAS: 80565-30-6 | C13H9ClO | 216.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 450360050
Price (EUR)
473,45
Online Exclusive
557,00
Save 83,55 (15%)
Each
-
Add to cart
Quantity:
5 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
473,45
Online Exclusive
557,00
Save 83,55 (15%)
Each
Add to cart
Chemical Identifiers
CAS80565-30-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting point112°C to 118°C
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
340572Certificate of AnalysisDec 12, 2022450360010, 450360050
344695Certificate of AnalysisOct 15, 2021450360010, 450360050
2 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Acute dermal toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P312 - Call a POISON CENTER or doctor if you feel unwell

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P337 + P313 - If eye irritation persists: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection