Tetrahydrofuran, 99.8%, for biochemistry, unstabilized, AcroSeal™
Tetrahydrofuran, 99.8%, for biochemistry, unstabilized, AcroSeal™
Tetrahydrofuran, 99.8%, for biochemistry, unstabilized, AcroSeal™
Tetrahydrofuran, 99.8%, for biochemistry, unstabilized, AcroSeal™
Thermo Scientific Chemicals

Tetrahydrofuran, 99.8%, for biochemistry, unstabilized, AcroSeal™

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 450530010
Price (USD)
89.65
Online Exclusive
99.70
Save 10.05 (10%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
89.65
Online Exclusive
99.70
Save 10.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Acidity=<0.0002 meq/g
Alkalinity=<0.0006 meq/g
Residue after evaporation=<2 ppm
UVat 230 nm A: =<0.5
UVat 240 nm A: =<0.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link